Product Details

SNP ID
rs147476338
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:114707917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAGTGAGGAGGTAGTTATTCTG[A/T]GAACTTCTCAAATAGAGATTATTGT
Phenotype
MIM: 191510 MIM: 164790
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CSDE1 PubMed Links

Gene Details

Gene
CSDE1
Gene Name
cold shock domain containing E1
There are no transcripts associated with this gene.

Gene
NRAS
Gene Name
neuroblastoma RAS viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002524.4 1001 UTR 3 NP_002515.1

View Full Product Details