Product Details

SNP ID
rs147508288
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:158398617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTGTAGACAAGAGGATTAAGTA[A/T]GGGAGTAACCACTGTGTAGGTCACT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10T2 PubMed Links

Gene Details

Gene
OR10T2
Gene Name
olfactory receptor family 10 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004475.1 850 Missense Mutation ATA,TTA I284L NP_001004475.1

View Full Product Details