Product Details

SNP ID
rs147825135
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:160371116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGCTATATCTCCTACCTGAAC[C/T]ACGTGCTGGACGTCTGAACTCAGCC
Phenotype
MIM: 162360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NHLH1 PubMed Links

Gene Details

Gene
NHLH1
Gene Name
nescient helix-loop-helix 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005598.3 827 Missense Mutation CAC,TAC H129Y NP_005589.1

View Full Product Details