Product Details

SNP ID
rs147878262
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:25834231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTCTCTCCACACCTGGGTTGT[A/G]GGGAGATTTGCTAATCACTGAAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AUNIP PubMed Links

Gene Details

Gene
AUNIP
Gene Name
aurora kinase A and ninein interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287490.1 1937 Intron NP_001274419.1
NM_024037.2 1937 UTR 3 NP_076942.1
XM_017002300.1 1937 UTR 3 XP_016857789.1
Gene
MTFR1L
Gene Name
mitochondrial fission regulator 1 like
There are no transcripts associated with this gene.

View Full Product Details