Product Details

SNP ID
rs147930858
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:175323409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCCATGAGACGGTGGTTGTAG[A/G]GGCGCATCTTCATTTCCACAAAGGG
Phenotype
MIM: 601995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNR PubMed Links

Gene Details

Gene
TNR
Gene Name
tenascin R
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003285.2 4189 Missense Mutation CCC,CTC P1342L NP_003276.3
XM_011509949.2 4189 Missense Mutation CCC,CTC P1183L XP_011508251.1
XM_017002218.1 4189 Missense Mutation CCC,CTC P1342L XP_016857707.1
XM_017002219.1 4189 Missense Mutation CCC,CTC P1342L XP_016857708.1

View Full Product Details