Product Details
- SNP ID
-
rs148033236
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:158254651 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGACCAATTTTTCTGAGAGAAGGA[A/G]ATAACATCTGCAAATGATATGCTGT
- Phenotype
-
MIM: 188370
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD1A
PubMed Links
Gene Details
- Gene
- CD1A
- Gene Name
- CD1a molecule
View Full Product Details