Product Details

SNP ID
rs148082143
Assay Type
Functionally Tested
NCBI dbSNP Submissions
9
Location
Chr.1:202594299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCACCAGAGGTTCCTACCATTGC[C/G]TTTCCTCCCTGTCAAGGGAATGCAA
Phenotype
MIM: 603768 MIM: 600104
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PPP1R12B PubMed Links

Gene Details

Gene
PPP1R12B
Gene Name
protein phosphatase 1 regulatory subunit 12B
There are no transcripts associated with this gene.

Gene
SYT2
Gene Name
synaptotagmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136504.1 4575 UTR 3 NP_001129976.1
NM_177402.4 4575 UTR 3 NP_796376.2
XM_011509192.2 4575 UTR 3 XP_011507494.1
XM_017000309.1 4575 UTR 3 XP_016855798.1
XM_017000310.1 4575 UTR 3 XP_016855799.1
XM_017000311.1 4575 UTR 3 XP_016855800.1
XM_017000312.1 4575 UTR 3 XP_016855801.1
XM_017000313.1 4575 UTR 3 XP_016855802.1

View Full Product Details