Product Details

SNP ID
rs148121416
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:21816930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTTTGAACTTCAGCCACATAA[C/T]CTCCTCCCTGCTCAGCATCTCTGCG
Phenotype
MIM: 142461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPG2 PubMed Links

Gene Details

Gene
HSPG2
Gene Name
heparan sulfate proteoglycan 2
There are no transcripts associated with this gene.

Gene
LDLRAD2
Gene Name
low density lipoprotein receptor class A domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013693.2 Intron NP_001013715.2
XM_005245873.4 Intron XP_005245930.1

View Full Product Details