Product Details
- SNP ID
-
hCV166817255
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:147640785 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTACATTTTATTTAAAGGCCAAAC[A/G]GAGAGCGGAATCCTGAGAAGACCAG
- Phenotype
-
MIM: 611471
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACP6
PubMed Links
Gene Details
- Gene
- ACP6
- Gene Name
- acid phosphatase 6, lysophosphatidic
There are no transcripts associated with this gene.
View Full Product Details