Product Details

SNP ID
rs148140635
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:248406122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCATCTTATTTGGAAGATATTAC[C/G]TTATATCGGCACAACTGTAGGATCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2T1 PubMed Links

Gene Details

Gene
OR2T1
Gene Name
olfactory receptor family 2 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030904.1 128 Missense Mutation CCT,CGT P43R NP_112166.1

View Full Product Details