Product Details

SNP ID
rs148156721
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:87332034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGACCATGGTGAATCCGGGCAGC[A/T]GCTCGCAGCCGCCCCCGGTGACGGC
Phenotype
MIM: 603129
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LMO4 PubMed Links

Gene Details

Gene
LMO4
Gene Name
LIM domain only 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006769.3 2969 Missense Mutation AGC,TGC S7C NP_006760.1
XM_005271291.3 2969 Missense Mutation AGC,TGC S7C XP_005271348.1

View Full Product Details