Product Details

SNP ID
rs148280953
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:204201787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTCTTCCTCAGGCCAATGATGA[A/G]GGACAGGCCCGTCAGGAACAGCAGC
Phenotype
MIM: 603286
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLT1A PubMed Links

Gene Details

Gene
GOLT1A
Gene Name
golgi transport 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198447.1 328 Missense Mutation CTC,TTC L48F NP_940849.1
XM_017000314.1 328 Missense Mutation CTC,TTC L48F XP_016855803.1
Gene
KISS1
Gene Name
KiSS-1 metastasis-suppressor
There are no transcripts associated with this gene.

View Full Product Details