Product Details

SNP ID
rs148390696
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:156292816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATGCAAGAAGGCAACACCAAGG[C/G]GACCTTATCAATAGCCCTGTCCCCT
Phenotype
MIM: 610962 MIM: 615531
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLMP PubMed Links

Gene Details

Gene
GLMP
Gene Name
glycosylated lysosomal membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256604.1 1152 UTR 3 NP_001243533.1
NM_001256605.1 1152 UTR 3 NP_001243534.1
NM_001256608.1 1152 UTR 3 NP_001243537.1
NM_001256609.1 1152 UTR 3 NP_001243538.1
NM_144580.2 1152 UTR 3 NP_653181.1
XM_011509117.2 1152 UTR 3 XP_011507419.1
XM_017000149.1 1152 UTR 3 XP_016855638.1
Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

Gene
TMEM79
Gene Name
transmembrane protein 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032323.2 1152 Intron NP_115699.1
Gene
VHLL
Gene Name
von Hippel-Lindau tumor suppressor like
There are no transcripts associated with this gene.

View Full Product Details