Product Details

SNP ID
rs148454740
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:15410270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGATCAAGGACATGGAGAAGA[G/T]GTTCAAGCAGTAAGTGCCCGCGCGA
Phenotype
MIM: 616450
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFHD2 PubMed Links

Gene Details

Gene
EFHD2
Gene Name
EF-hand domain family member D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024329.5 374 Missense Mutation AGG,ATG R100M NP_077305.2
XM_005246000.3 374 Missense Mutation AGG,ATG R100M XP_005246057.1
Gene
FHAD1
Gene Name
forkhead associated phosphopeptide binding domain 1
There are no transcripts associated with this gene.

Gene
LOC101927441
Gene Name
uncharacterized LOC101927441
There are no transcripts associated with this gene.

View Full Product Details