Product Details

SNP ID
rs148475750
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:163326152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATGGTAAAAACCTCACCAAGA[A/T]TGATCTTTATCCAAATCCAAAGGTA
Phenotype
MIM: 611772 MIM: 603276
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NUF2 PubMed Links

Gene Details

Gene
NUF2
Gene Name
NUF2, NDC80 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031423.3 401 Missense Mutation AAT,ATT N34I NP_113611.2
NM_145697.2 401 Missense Mutation AAT,ATT N34I NP_663735.2
XM_011510036.1 401 UTR 5 XP_011508338.1
Gene
RGS5
Gene Name
regulator of G-protein signaling 5
There are no transcripts associated with this gene.

View Full Product Details