Product Details

SNP ID
rs148510143
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:8355448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATCTTCCTGACTTGGTAGGTG[A/G]CCACCATGCATGTGGGGGTGTCCAT
Phenotype
MIM: 605226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RERE PubMed Links

Gene Details

Gene
RERE
Gene Name
arginine-glutamic acid dipeptide repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042681.1 5188 Silent Mutation GGC,GGT G1546G NP_001036146.1
NM_001042682.1 5188 Silent Mutation GGC,GGT G992G NP_001036147.1
NM_012102.3 5188 Silent Mutation GGC,GGT G1546G NP_036234.3
XM_005263464.2 5188 Silent Mutation GGC,GGT G1546G XP_005263521.1
XM_005263466.1 5188 Silent Mutation GGC,GGT G1278G XP_005263523.1
XM_011541510.1 5188 Silent Mutation GGC,GGT G1504G XP_011539812.1
XM_011541511.1 5188 Intron XP_011539813.1
XM_017001358.1 5188 Silent Mutation GGC,GGT G1546G XP_016856847.1
XM_017001359.1 5188 Silent Mutation GGC,GGT G1546G XP_016856848.1

View Full Product Details