Product Details

SNP ID
rs148588192
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:2592718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCAAGGGTGTTGAATCCGTT[C/G]ACCTGCGCACAGGAGACAGGATTGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM213B PubMed Links

Gene Details

Gene
FAM213B
Gene Name
family with sequence similarity 213 member B
There are no transcripts associated with this gene.

Gene
LOC100996583
Gene Name
uncharacterized LOC100996583
There are no transcripts associated with this gene.

Gene
MMEL1
Gene Name
membrane metallo-endopeptidase-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033467.3 2484 Silent Mutation GTC,GTG V668V NP_258428.2
XM_011542122.2 2484 Silent Mutation GTC,GTG V411V XP_011540424.1
XM_017002310.1 2484 Silent Mutation GTC,GTG V668V XP_016857799.1
XM_017002311.1 2484 Silent Mutation GTC,GTG V650V XP_016857800.1
XM_017002312.1 2484 Silent Mutation GTC,GTG V642V XP_016857801.1
XM_017002313.1 2484 Silent Mutation GTC,GTG V668V XP_016857802.1
XM_017002314.1 2484 Silent Mutation GTC,GTG V625V XP_016857803.1
XM_017002315.1 2484 Silent Mutation GTC,GTG V564V XP_016857804.1

View Full Product Details