Product Details

SNP ID
rs148947714
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:150226728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTCCTCTTCCTCTTCCTCCTC[C/T]TCTTCCTCATATCCTTCCGGTGGAC
Phenotype
MIM: 609611
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANP32E PubMed Links

Gene Details

Gene
ANP32E
Gene Name
acidic nuclear phosphoprotein 32 family member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136478.3 765 Silent Mutation GAA,GAG E146E NP_001129950.1
NM_001136479.2 765 Silent Mutation GAA,GAG E139E NP_001129951.1
NM_001280559.1 765 Silent Mutation GAA,GAG E187E NP_001267488.1
NM_001280560.1 765 Missense Mutation AAG,AGG K132R NP_001267489.1
NM_030920.4 765 Silent Mutation GAA,GAG E187E NP_112182.1
XM_005245513.3 765 Silent Mutation GAA,GAG E187E XP_005245570.1
XM_005245514.3 765 Silent Mutation GAA,GAG E187E XP_005245571.1
XM_017002418.1 765 Silent Mutation GAA,GAG E187E XP_016857907.1

View Full Product Details