Product Details

SNP ID
rs149024884
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:94982844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATACATATTTTTATCTAGAAAAG[A/C]AATTTGGTTTACAGAGGCATAAACA
Phenotype
MIM: 612866
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALG14 PubMed Links

Gene Details

Gene
ALG14
Gene Name
ALG14, UDP-N-acetylglucosaminyltransferase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305242.1 936 UTR 3 NP_001292171.1
NM_144988.3 936 UTR 3 NP_659425.1
XM_005270582.3 936 Intron XP_005270639.1
XM_011540897.2 936 Intron XP_011539199.1

View Full Product Details