Product Details

SNP ID
rs149027264
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:100348917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAAATTTTGTCATTTTCAGGGC[A/G]AGTGCAGTGGCTCATATCTGTAATT
Phenotype
MIM: 603504
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC14A PubMed Links

Gene Details

Gene
CDC14A
Gene Name
cell division cycle 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319210.1 Intron NP_001306139.1
NM_001319211.1 Intron NP_001306140.1
NM_001319212.1 Intron NP_001306141.1
NM_003672.3 Intron NP_003663.2
NM_033312.2 Intron NP_201569.1
NM_033313.2 Intron NP_201570.1
XM_005271294.3 Intron XP_005271351.1
XM_005271296.3 Intron XP_005271353.1
XM_011542340.2 Intron XP_011540642.1
XM_011542341.2 Intron XP_011540643.1
XM_011542345.2 Intron XP_011540647.1
XM_011542346.2 Intron XP_011540648.1
XM_017002646.1 Intron XP_016858135.1
XM_017002647.1 Intron XP_016858136.1

View Full Product Details