Product Details

SNP ID
rs149042704
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:247941620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGTGCTATTTGGACAGACGCC[A/G]AATGGAGAGAGATAGAGAAAAAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2L13 PubMed Links

Gene Details

Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 Intron NP_001291464.1
NM_175911.3 Intron NP_787107.1
XM_011544169.2 Intron XP_011542471.1
Gene
OR2L8
Gene Name
olfactory receptor family 2 subfamily L member 8 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details