Product Details
- SNP ID
-
rs149108666
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:42153462 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCAGGGAGCGCGATGGGCTGCAG[A/G]GCTGCATCAGGGCTCCTGCCAGGAG
- Phenotype
-
MIM: 139392
MIM: 601271
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GUCA2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1047047] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GUCA2A
- Gene Name
- guanylate cyclase activator 2A
There are no transcripts associated with this gene.
- Gene
- GUCA2B
- Gene Name
- guanylate cyclase activator 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007102.2 |
42 |
Silent Mutation |
AGA,AGG |
R4R |
NP_009033.1 |
View Full Product Details