Product Details
- SNP ID
-
rs149177918
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:20589258 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTTCCTGTGGGGGCTGCCCTGCT[C/T]ACCCAGGAGGGGAGAATCTTCAAAG
- Phenotype
-
MIM: 123920
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDA
PubMed Links
Gene Details
- Gene
- CDA
- Gene Name
- cytidine deaminase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001785.2 |
308 |
Silent Mutation |
CTC,CTT |
L43L |
NP_001776.1 |
View Full Product Details