Product Details

SNP ID
rs149283693
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:155942117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCTTCCTCATCACAGCGCTGAG[C/T]GTTGCTCGCTACTGGGTGGTGGCCA
Phenotype
MIM: 607560 MIM: 609043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF2 PubMed Links

Gene Details

Gene
ARHGEF2
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
There are no transcripts associated with this gene.

Gene
KIAA0907
Gene Name
KIAA0907
There are no transcripts associated with this gene.

Gene
MIR6738
Gene Name
microRNA 6738
There are no transcripts associated with this gene.

Gene
RXFP4
Gene Name
relaxin/insulin like family peptide receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181885.2 429 Silent Mutation AGC,AGT S136S NP_871001.1

View Full Product Details