Product Details

SNP ID
rs149351367
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:29148453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGATCTAGATCGGGAGGGCGAT[C/G]TGGAAGCAGACCTGGATCTAGACTT
Phenotype
MIM: 601940
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SRSF4 PubMed Links

Gene Details

Gene
SRSF4
Gene Name
serine and arginine rich splicing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005626.4 1525 Missense Mutation ACA,AGA T481R NP_005617.2
XM_011541951.2 1525 Missense Mutation ACA,AGA T451R XP_011540253.1

View Full Product Details