Product Details

SNP ID
rs149353254
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:220102216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCAGATTGGAATAATTGCTACT[A/G]TACATTTGATGGGAAGTATGAAGCC
Phenotype
MIM: 612801
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IARS2 PubMed Links

Gene Details

Gene
IARS2
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018060.3 742 Missense Mutation TAT,TGT Y213C NP_060530.3

View Full Product Details