Product Details

SNP ID
rs149604328
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:168581873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGAAACTTTATTCAAGTAAAA[A/G]TCAATACCCTTTGAATTGGACAATA
Phenotype
MIM: 600250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XCL1 PubMed Links

Gene Details

Gene
XCL1
Gene Name
X-C motif chemokine ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002995.2 1163 UTR 3 NP_002986.1
XM_011509865.2 1163 Intron XP_011508167.1

View Full Product Details