Product Details

SNP ID
rs149832483
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:160040232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTCTGCTCCTGCCCAAACCTTA[A/G]CAAAGCATCACCAATTCTCAGAGGC
Phenotype
MIM: 602208 MIM: 610273
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNJ10 PubMed Links

Gene Details

Gene
KCNJ10
Gene Name
potassium voltage-gated channel subfamily J member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002241.4 2541 UTR 3 NP_002232.2
Gene
PIGM
Gene Name
phosphatidylinositol glycan anchor biosynthesis class M
There are no transcripts associated with this gene.

View Full Product Details