Product Details

SNP ID
rs149965873
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:26235090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTGGTTGAACTGAAAGTTAGT[A/G]TGTTCAGGTTTATTTGTTCAACAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP85 PubMed Links

Gene Details

Gene
CEP85
Gene Name
centrosomal protein 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281517.2 Intron NP_001268446.1
NM_001281518.2 Intron NP_001268447.1
NM_001319944.1 Intron NP_001306873.1
NM_022778.4 Intron NP_073615.2
XM_017002105.1 Intron XP_016857594.1
Gene
LOC101928303
Gene Name
uncharacterized LOC101928303
There are no transcripts associated with this gene.

View Full Product Details