Product Details

SNP ID
rs149978254
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:116579066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTAATTCGCTAAAGACTAGTTCC[A/G]ATATTCTAAAAAATAACAAAGTAAC
Phenotype
MIM: 153420 MIM: 603491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD58 PubMed Links

Gene Details

Gene
CD58
Gene Name
CD58 molecule
There are no transcripts associated with this gene.

Gene
IGSF3
Gene Name
immunoglobulin superfamily member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007237.2 Intron NP_001007238.1
NM_001542.3 Intron NP_001533.2
XM_005270793.2 Intron XP_005270850.1
XM_005270794.4 Intron XP_005270851.1
XM_006710593.3 Intron XP_006710656.1
XM_011541315.1 Intron XP_011539617.1
XM_011541316.2 Intron XP_011539618.1

View Full Product Details