Product Details

SNP ID
rs149995860
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:12234363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTCTCAGTTGCACTTCTCAAA[C/G]GTGAGTATTTCTCTGGGTGAGATAC
Phenotype
MIM: 608877
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VPS13D PubMed Links

Gene Details

Gene
VPS13D
Gene Name
vacuolar protein sorting 13 homolog D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015378.3 255 Missense Mutation CGT,GGT R33G NP_056193.2
NM_018156.3 255 Missense Mutation CGT,GGT R33G NP_060626.2

View Full Product Details