Product Details

SNP ID
rs150046345
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:153776089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGCCTGCGCACTGCCTTTGTG[C/T]CCACCGCCCTGCGCCGGGGCCCCCT
Phenotype
MIM: 611347 MIM: 604193
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INTS3 PubMed Links

Gene Details

Gene
INTS3
Gene Name
integrator complex subunit 3
There are no transcripts associated with this gene.

Gene
SLC27A3
Gene Name
solute carrier family 27 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317929.1 798 Missense Mutation CCC,TCC P245S NP_001304858.1
NM_024330.1 798 Missense Mutation CCC,TCC P245S NP_077306.1

View Full Product Details