Product Details

SNP ID
rs150535630
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:228174815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGTCACGGTGGAGCCGCACCC[G/T]GAGCTGCGAGTGTGGGCGGTGTTGC
Phenotype
MIM: 615316
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IBA57 PubMed Links

Gene Details

Gene
IBA57
Gene Name
IBA57 homolog, iron-sulfur cluster assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010867.3 474 Silent Mutation CCG,CCT P155P NP_001010867.1
NM_001310327.1 474 UTR 5 NP_001297256.1
Gene
IBA57-AS1
Gene Name
IBA57 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details