Product Details

SNP ID
rs150537161
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:32894811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCAGGACATCATCTCTCAGAG[C/G]GTCAGAGGCTCAGAGCTGGTGCAGC
Phenotype
MIM: 142622
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HPCA PubMed Links

Gene Details

Gene
HPCA
Gene Name
hippocalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002143.2 914 Intron NP_002134.2
XM_005270792.2 914 Intron XP_005270849.1
XM_017001118.1 914 Intron XP_016856607.1
Gene
TMEM54
Gene Name
transmembrane protein 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005270408.3 914 Silent Mutation ACC,ACG T239T XP_005270465.1

View Full Product Details