Product Details

SNP ID
rs150575664
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:154965671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCATTGAGCTGCAGCAGTGCCT[C/T]AGCCTCCCGCCGGCTCAGCTTCCCA
Phenotype
MIM: 116900 MIM: 606903 MIM: 600560
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CKS1B PubMed Links

Gene Details

Gene
CKS1B
Gene Name
CDC28 protein kinase regulatory subunit 1B
There are no transcripts associated with this gene.

Gene
LOC101928120
Gene Name
uncharacterized LOC101928120
There are no transcripts associated with this gene.

Gene
PBXIP1
Gene Name
PBX homeobox interacting protein 1
There are no transcripts associated with this gene.

Gene
PYGO2
Gene Name
pygopus family PHD finger 2
There are no transcripts associated with this gene.

Gene
SHC1
Gene Name
SHC adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130040.1 1245 Intron NP_001123512.1
NM_001130041.1 1245 Intron NP_001123513.1
NM_001202859.1 1245 Intron NP_001189788.1
NM_003029.4 1245 Intron NP_003020.2
NM_183001.4 1245 Intron NP_892113.4
XM_005245449.4 1245 Missense Mutation AAG,GAG K499E XP_005245506.1
XM_005245451.4 1245 Missense Mutation AAG,GAG K389E XP_005245508.1
XM_011509892.2 1245 Missense Mutation AAG,GAG K509E XP_011508194.1
XM_011509893.2 1245 Missense Mutation AAG,GAG K508E XP_011508195.1
XM_011509894.2 1245 Missense Mutation AAG,GAG K491E XP_011508196.1
XM_011509897.1 1245 Intron XP_011508199.1
XM_011509898.2 1245 Missense Mutation AAG,GAG K333E XP_011508200.1
XM_017002081.1 1245 Missense Mutation AAG,GAG K482E XP_016857570.1
XM_017002082.1 1245 Missense Mutation AAG,GAG K481E XP_016857571.1
XM_017002083.1 1245 Intron XP_016857572.1

View Full Product Details