Product Details

SNP ID
rs150601677
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:248626334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGACACTACTTTGTCCTGCTC[A/G]GGGGTGTGGAAGGACTGGGGCAGCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR2T10 PubMed Links

Gene Details

Gene
OR2T10
Gene Name
olfactory receptor family 2 subfamily T member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004693.1 795 Intron NP_001004693.1
XM_017000273.1 795 Intron XP_016855762.1
Gene
OR2T11
Gene Name
olfactory receptor family 2 subfamily T member 11 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001964.1 795 Silent Mutation NP_001001964.1

View Full Product Details