Product Details

SNP ID
rs150710676
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:21815994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATGGCAGGTGCATCCCCTCAA[A/G]CCTCGTGTGTGACCCCTGGGGCATG
Phenotype
MIM: 142461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPG2 PubMed Links

Gene Details

Gene
HSPG2
Gene Name
heparan sulfate proteoglycan 2
There are no transcripts associated with this gene.

Gene
LDLRAD2
Gene Name
low density lipoprotein receptor class A domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013693.2 750 Missense Mutation AAC,AGC N188S NP_001013715.2
XM_005245873.4 750 Missense Mutation AAC,AGC N188S XP_005245930.1

View Full Product Details