Product Details

SNP ID
rs150717400
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:158466159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGGCTTCAGTCAGCTGGTCATA[G/T]TCATGCTTGGTGTATTTGCCTTGGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10K1 PubMed Links

Gene Details

Gene
OR10K1
Gene Name
olfactory receptor family 10 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004473.1 598 Missense Mutation GTC,TTC V200F NP_001004473.1

View Full Product Details