Product Details

SNP ID
rs150742629
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:15942095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGCACCTGCCCGGCAGGCCA[C/T]GTGCCACCTGGCCCATACTGCTGAT
Phenotype
MIM: 613484 MIM: 604084
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPEN PubMed Links

Gene Details

Gene
SPEN
Gene Name
spen family transcriptional repressor
There are no transcripts associated with this gene.

Gene
ZBTB17
Gene Name
zinc finger and BTB domain containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242884.1 2513 Silent Mutation NP_001229813.1
NM_001287603.1 2513 Silent Mutation NP_001274532.1
NM_001287604.1 2513 Silent Mutation NP_001274533.1
NM_001324137.1 2513 Silent Mutation NP_001311066.1
NM_001324138.1 2513 Silent Mutation NP_001311067.1
NM_003443.2 2513 Silent Mutation NP_003434.2
XM_005245986.1 2513 Silent Mutation XP_005246043.1
XM_005245987.1 2513 Silent Mutation XP_005246044.1
XM_011542085.2 2513 Silent Mutation XP_011540387.1
XM_011542086.1 2513 Silent Mutation XP_011540388.1
XM_011542087.2 2513 Silent Mutation XP_011540389.1
XM_011542088.1 2513 Silent Mutation XP_011540390.1
XM_017002242.1 2513 Silent Mutation XP_016857731.1

View Full Product Details