Product Details

SNP ID
rs150747052
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:158579249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCAGCAAACCAGACACTGAGAT[C/G]AGTGTTATAATGAATTCTGTGTGGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10X1 PubMed Links

Gene Details

Gene
OR10X1
Gene Name
olfactory receptor family 10 subfamily X member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004477.1 651 Silent Mutation CTC,CTG L217L NP_001004477.1

View Full Product Details