Product Details

SNP ID
rs150910351
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:111147773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATTCTCACCCGGAGTCCCCAG[C/T]GGGCTTCGGGCATATGAGTACTACA
Phenotype
MIM: 616751 MIM: 613360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEPT1 PubMed Links

Gene Details

Gene
CEPT1
Gene Name
choline/ethanolamine phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006710285.3 229 Missense Mutation GCG,GTG A20V XP_006710348.1
XM_011540496.1 229 Missense Mutation GCG,GTG A20V XP_011538798.1
Gene
DRAM2
Gene Name
DNA damage regulated autophagy modulator 2
There are no transcripts associated with this gene.

View Full Product Details