Product Details

SNP ID
rs151092831
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:39659251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATGGAACATCTGGTCATCAAAG[A/T]AGATGTGTGGGCGGATCTTCTCAAG
Phenotype
MIM: 610525
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NT5C1A PubMed Links

Gene Details

Gene
NT5C1A
Gene Name
5'-nucleotidase, cytosolic IA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032526.2 977 Missense Mutation TAC,TTC Y326F NP_115915.1

View Full Product Details