Product Details

SNP ID
rs151238352
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:943346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATGACGTCTGCAGCTTCGTGG[A/G]GGGCCTGTCTGGCTGTGGAGAGTAC
Phenotype
MIM: 610770 MIM: 616765
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOC2L PubMed Links

Gene Details

Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015658.3 1738 Intron NP_056473.2
Gene
SAMD11
Gene Name
sterile alpha motif domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152486.2 1738 Missense Mutation GAG,GGG E553G NP_689699.2

View Full Product Details