Product Details

SNP ID
rs151318203
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:158547391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGGTGGAGAATGCCTTTTGGC[A/G]GCCCTGAGCAGAAGGGATCCTGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6Y1 PubMed Links

Gene Details

Gene
OR6Y1
Gene Name
olfactory receptor family 6 subfamily Y member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005189.1 715 Missense Mutation CGC,TGC R239C NP_001005189.1

View Full Product Details