Product Details

SNP ID
rs138084629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1570992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCATCCTTGTTCTCTATGAGGGT[C/T]GAAGCTGTTTCTGTCCGGGACACAT
Phenotype
MIM: 603889
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIRPB1 PubMed Links

Gene Details

Gene
SIRPB1
Gene Name
signal regulatory protein beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005260641.3 1400 Silent Mutation TCA,TCG S298S XP_005260698.1
XM_005260643.3 1400 Intron XP_005260700.1
XM_005260644.3 1400 Intron XP_005260701.1
XM_011529130.2 1400 Intron XP_011527432.1
XM_011529134.2 1400 Intron XP_011527436.1
XM_017027577.1 1400 Silent Mutation TCA,TCG S299S XP_016883066.1
XM_017027578.1 1400 Silent Mutation TCA,TCG S299S XP_016883067.1
XM_017027579.1 1400 Silent Mutation TCA,TCG S299S XP_016883068.1
XM_017027580.1 1400 Silent Mutation TCA,TCG S299S XP_016883069.1
XM_017027581.1 1400 Intron XP_016883070.1
XM_017027582.1 1400 Intron XP_016883071.1
XM_017027583.1 1400 Silent Mutation TCA,TCG S299S XP_016883072.1
XM_017027584.1 1400 Silent Mutation TCA,TCG S299S XP_016883073.1
XM_017027585.1 1400 Intron XP_016883074.1

View Full Product Details