Product Details

SNP ID
rs138752668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45886793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCCCACTGCCTCAGAGGGTAC[C/T]GGTAGAACCCGTGGTGACAGCCCAC
Phenotype
MIM: 613111 MIM: 608597
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSA PubMed Links

Gene Details

Gene
CTSA
Gene Name
cathepsin A
There are no transcripts associated with this gene.

Gene
NEURL2
Gene Name
neuralized E3 ubiquitin protein ligase 2
There are no transcripts associated with this gene.

Gene
SPATA25
Gene Name
spermatogenesis associated 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080608.3 417 Silent Mutation CCA,CCG P136P NP_542175.1
Gene
ZSWIM1
Gene Name
zinc finger SWIM-type containing 1
There are no transcripts associated with this gene.

Gene
ZSWIM3
Gene Name
zinc finger SWIM-type containing 3
There are no transcripts associated with this gene.

View Full Product Details