Product Details

SNP ID
rs139432314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:48626334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGACCAGACAGGAGGCTGCGTG[A/G]TGTCCAGGGCAAGCAGACTGCAATG
Phenotype
MIM: 606905
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PREX1 PubMed Links

Gene Details

Gene
PREX1
Gene Name
phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020820.3 Intron NP_065871.2
XM_011528934.2 Intron XP_011527236.1

View Full Product Details