Product Details
- SNP ID
-
rs141459148
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31968485 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCGTGCTCCTGCCCTCGCTGGTC[C/G]TGCAGTTACTGAGCTTCCGCTGGTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
XKR7
PubMed Links
Gene Details
- Gene
- XKR7
- Gene Name
- XK related 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001011718.1 |
484 |
Missense Mutation |
CTG,GTG |
L104V |
NP_001011718.1 |
View Full Product Details