Product Details

SNP ID
rs141746104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37254301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTACCTGACGACCAAGCCGTG[C/T]CCTTGCTCCTCGCTCTTGGTTGCTC
Phenotype
MIM: 139190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GHRH PubMed Links

Gene Details

Gene
GHRH
Gene Name
growth hormone releasing hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184731.2 236 Missense Mutation ACA,GCA T73A NP_001171660.1
NM_021081.5 236 Missense Mutation ACA,GCA T73A NP_066567.1
XM_011528784.2 236 Missense Mutation ACA,GCA T73A XP_011527086.1
XM_011528788.2 236 Missense Mutation ACA,GCA T73A XP_011527090.1

View Full Product Details