Product Details

SNP ID
rs142460450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36240118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTTCTTCCTTAGCCTGCACCAG[C/T]GGGGGCTGACAGTGCTGCGCTGGAG
Phenotype
MIM: 602879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AAR2 PubMed Links

Gene Details

Gene
AAR2
Gene Name
AAR2 splicing factor homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271874.1 475 Missense Mutation CGG,TGG R84W NP_001258803.1
NM_015511.4 475 Missense Mutation CGG,TGG R84W NP_056326.2
XM_006723770.3 475 Missense Mutation CGG,TGG R84W XP_006723833.1
XM_011528762.2 475 Missense Mutation CGG,TGG R84W XP_011527064.1
XM_011528763.2 475 Missense Mutation CGG,TGG R84W XP_011527065.1
Gene
EPB41L1
Gene Name
erythrocyte membrane protein band 4.1 like 1
There are no transcripts associated with this gene.

View Full Product Details